Assay Details
| Gene Symbol: Snx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858141.3 | XP_004858198.1 | 1 - 2 | 420 | 80 | |
| XM_021239007.1 | XP_021094666.1 | 1 - 2 | 334 | 80 | ||
More Information
Set Membership: |
|

Snx21
Genomic Map