Assay Details
| Gene Symbol: Ltb4r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864087.3 | XP_004864144.1 | 1 - 2 | 168 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004864087.3 transcripts.
Set Membership: |
|

Ltb4r2
Genomic Map