Assay Details
| Gene Symbol: Anp32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864954.3 | XP_004865011.1 | 4 - 5 | 831 | 72 | |
| XM_021244500.1 | XP_021100159.1 | 5 - 6 | 878 | 72 | ||
| XM_021244501.1 | XP_021100160.1 | 4 - 5 | 545 | 72 | ||
More Information
Set Membership: |
|

Anp32b
Genomic Map