Assay Details
| Gene Symbol: Lzts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866331.3 | XP_004866388.1 | 2 - 3 | 551 | 61 | |
| XM_004866332.3 | XP_004866389.1 | 2 - 3 | 599 | 61 | ||
| XM_004866333.3 | XP_004866390.1 | 2 - 3 | 524 | 61 | ||
| XM_021245627.1 | XP_021101286.1 | 2 - 3 | 525 | 61 | ||
| XM_021245628.1 | XP_021101287.1 | 3 - 4 | 462 | 61 | ||
| XM_021245629.1 | XP_021101288.1 | 3 - 4 | 459 | 61 | ||
| XM_021245630.1 | XP_021101289.1 | 3 - 4 | 506 | 61 | ||
More Information
Set Membership: |
|

Lzts2
Genomic Map