Assay Details
| Gene Symbol: Vsig2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874465.3 | XP_004874522.1 | 1 - 2 | 473 | 78 | |
| XM_004874467.3 | XP_004874524.1 | 3 - 4 | 447 | 78 | ||
| XM_021251904.1 | XP_021107563.1 | 2 - 3 | 773 | 78 | ||
More Information
Set Membership: |
|

Vsig2
Genomic Map