Assay Details
| Gene Symbol: Galnt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021244512.1 | XP_021100171.1 | 10 - 11 | 1737 | 69 | |
| XM_021244513.1 | XP_021100172.1 | 9 - 10 | 1649 | 69 | ||
More Information
Set Membership: |
|

Galnt12
Genomic Map