Assay Details
| Gene Symbol: Usf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835704.2 | XP_004835761.1 | 2 - 3 | 401 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004835704.2 transcripts.
Set Membership: |
|

Usf3
Genomic Map