Assay Details
| Gene Symbol: Usf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835704.2 | XP_004835761.1 | 5 - 6 | 579 | 98 | |
| XM_004835705.2 | XP_004835762.1 | 4 - 5 | 454 | 98 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usf3| Gene Symbol: Usf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835704.2 | XP_004835761.1 | 5 - 6 | 579 | 98 | |
| XM_004835705.2 | XP_004835762.1 | 4 - 5 | 454 | 98 | ||