Assay Details
| Gene Symbol: Amn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836909.3 | XP_004836966.1 | 7 - 8 | 797 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amn| Gene Symbol: Amn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836909.3 | XP_004836966.1 | 7 - 8 | 797 | 56 | |
Set Membership: |
|