Assay Details
| Gene Symbol: Fbxo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844783.3 | XP_004844840.2 | 2 - 3 | 372 | 57 | |
| XM_004844784.3 | XP_004844841.1 | 2 - 3 | 590 | 57 | ||
| XM_004844786.3 | XP_004844843.1 | 2 - 3 | 220 | 57 | ||
| XM_021258344.1 | XP_021114003.1 | 2 - 3 | 593 | 57 | ||
| XM_021258345.1 | XP_021114004.1 | 2 - 3 | 200 | 57 | ||
More Information
Set Membership: |
|

Fbxo7
Genomic Map