Assay Details
| Gene Symbol: Mcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845444.2 | XP_004845501.1 | 2 - 3 | 1168 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcat| Gene Symbol: Mcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845444.2 | XP_004845501.1 | 2 - 3 | 1168 | 57 | |
Set Membership: |
|