Assay Details
| Gene Symbol: Bst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847508.2 | XP_004847565.1 | 2 - 3 | 381 | 65 | |
| XM_021260257.1 | XP_021115916.1 | 2 - 3 | 361 | 65 | ||
| XM_021260258.1 | XP_021115917.1 | 2 - 3 | 381 | 65 | ||
More Information
Set Membership: |
|

Bst1
Genomic Map