Assay Details
| Gene Symbol: Ccdc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851792.3 | XP_004851849.2 | 4 - 5 | 763 | 85 | |
| XM_021263943.1 | XP_021119602.1 | 4 - 5 | 763 | 85 | ||
| XM_021263944.1 | XP_021119603.1 | 4 - 5 | 763 | 85 | ||
| XM_021263945.1 | XP_021119604.1 | 4 - 5 | 763 | 85 | ||
| XM_021263946.1 | XP_021119605.1 | 4 - 5 | 763 | 85 | ||
| XM_021263947.1 | XP_021119606.1 | 4 - 5 | 763 | 85 | ||
| XM_021263948.1 | XP_021119607.1 | 3 - 4 | 690 | 85 | ||
More Information
Set Membership: |
|

Ccdc34
Genomic Map