Assay Details
| Gene Symbol: Vapa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853238.3 | XP_004853295.1 | 6 - 7 | 838 | 83 | |
| XM_004853239.3 | XP_004853296.1 | 5 - 6 | 703 | 83 | ||
More Information
Set Membership: |
|

Vapa
Genomic Map