Assay Details
| Gene Symbol: Cop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853391.3 | XP_004853448.1 | 13 - 14 | 1749 | 68 | |
| XM_013075379.2 | XP_012930833.1 | 13 - 14 | 1519 | 68 | ||
| XM_021265388.1 | XP_021121047.1 | 13 - 14 | 1531 | 68 | ||
More Information
Set Membership: |
|

Cop1
Genomic Map