Assay Details
| Gene Symbol: Lnx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854811.3 | XP_004854868.1 | 2 - 3 | 589 | 77 | |
| XM_013075856.2 | XP_012931310.1 | 2 - 3 | 589 | 77 | ||
| XM_013075857.2 | XP_012931311.1 | 2 - 3 | 670 | 77 | ||
More Information
Set Membership: |
|

Lnx2
Genomic Map