Assay Details
| Gene Symbol: Zbtb7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855114.2 | XP_004855171.1 | 3 - 4 | 463 | 91 | |
| XM_004855115.2 | XP_004855172.1 | 2 - 3 | 251 | 91 | ||
| XM_021236603.1 | XP_021092262.1 | 4 - 5 | 558 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004855114.2, XM_004855115.2, XM_021236603.1 transcripts.
Set Membership: |
|

Zbtb7c
Genomic Map