Assay Details
| Gene Symbol: Ttc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856712.3 | XP_004856769.1 | 1 - 2 | 220 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc36| Gene Symbol: Ttc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856712.3 | XP_004856769.1 | 1 - 2 | 220 | 77 | |
Set Membership: |
|