Assay Details
| Gene Symbol: Rflnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857108.2 | XP_004857165.1 | 2 - 3 | 450 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rflnb| Gene Symbol: Rflnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857108.2 | XP_004857165.1 | 2 - 3 | 450 | 74 | |
Set Membership: |
|