Assay Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860762.3 | XP_004860819.1 | 7 - 8 | 811 | 60 | |
| XM_021241311.1 | XP_021096970.1 | 5 - 6 | 804 | 60 | ||
More Information
Set Membership: |
|

St6galnac2
Genomic Map