Assay Details
| Gene Symbol: Scnn1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863936.3 | XP_004863993.1 | 9 - 10 | 1392 | 65 | |
| XM_021243623.1 | XP_021099282.1 | 9 - 10 | 1365 | 65 | ||
| XM_021243624.1 | XP_021099283.1 | 9 - 10 | 1392 | 65 | ||
More Information
Set Membership: |
|

Scnn1d
Genomic Map