Assay Details
| Gene Symbol: Rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869252.2 | XP_004869309.1 | 4 - 5 | 628 | 56 | |
| XM_004869253.3 | XP_004869310.1 | 4 - 5 | 608 | 56 | ||
| XM_021247966.1 | XP_021103625.1 | 4 - 5 | 669 | 56 | ||
More Information
Set Membership: |
|

Rab3b
Genomic Map