Assay Details
| Gene Symbol: Cpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875037.3 | XP_004875094.1 | 4 - 5 | 498 | 83 | |
| XM_021252361.1 | XP_021108020.1 | 4 - 5 | 498 | 83 | ||
| XM_021252362.1 | XP_021108021.1 | 4 - 5 | 498 | 83 | ||
More Information
Set Membership: |
|

Cpm
Genomic Map