Assay Details
| Gene Symbol: Atf7ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065617.2 | XP_012921071.1 | 7 - 8 | 1524 | 91 | |
| XM_013065618.2 | XP_012921072.1 | 7 - 8 | 1513 | 91 | ||
| XM_013065619.2 | XP_012921073.1 | 6 - 7 | 1425 | 91 | ||
| XM_013065620.2 | XP_012921074.1 | 6 - 7 | 1389 | 91 | ||
| XM_013065621.2 | XP_012921075.1 | 7 - 8 | 1524 | 91 | ||
| XM_013065622.2 | XP_012921076.1 | 5 - 6 | 1290 | 91 | ||
| XM_013065623.2 | XP_012921077.1 | 7 - 8 | 1524 | 91 | ||
| XM_013065624.2 | XP_012921078.1 | 7 - 8 | 1524 | 91 | ||
| XM_013065626.2 | XP_012921080.1 | 7 - 8 | 1524 | 91 | ||
| XM_021244478.1 | XP_021100137.1 | 8 - 9 | 1636 | 91 | ||
| XM_021244479.1 | XP_021100138.1 | 7 - 8 | 1534 | 91 | ||
| XM_021244480.1 | XP_021100139.1 | 7 - 8 | 1524 | 91 | ||
More Information
Set Membership: |
|

Atf7ip2
Genomic Map