Assay Details
| Gene Symbol: Znf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069019.2 | XP_012924473.1 | 2 - 3 | 684 | 101 | |
| XM_013069020.2 | XP_012924474.1 | 2 - 3 | 352 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013069019.2, XM_013069020.2 transcripts.
Set Membership: |
|

Znf25
Genomic Map