Assay Details
| Gene Symbol: Eef1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069444.1 | XP_012924898.1 | 2 - 3 | 1220 | 86 | |
| XM_013069448.2 | XP_012924902.1 | 2 - 3 | 196 | 86 | ||
| XM_013069452.2 | XP_012924906.1 | 2 - 3 | 264 | 86 | ||
| XM_021250924.1 | XP_021106583.1 | 2 - 3 | 1220 | 86 | ||
| XM_021250942.1 | XP_021106601.1 | 2 - 3 | 142 | 86 | ||
| XM_021250949.1 | XP_021106608.1 | 2 - 3 | 238 | 86 | ||
| XM_021250957.1 | XP_021106616.1 | 1 - 2 | 138 | 86 | ||
More Information
Set Membership: |
|

Eef1d
Genomic Map