Assay Details
| Gene Symbol: Gtsf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239069.1 | XP_021094728.1 | 1 | 623 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtsf1l| Gene Symbol: Gtsf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239069.1 | XP_021094728.1 | 1 | 623 | 76 | |
Set Membership: |
|