Assay Details
| Gene Symbol: Il7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255999.1 | XP_021111658.1 | 3 - 4 | 481 | 110 | |
| XM_021256000.1 | XP_021111659.1 | 3 - 4 | 809 | 110 | ||
| XM_021256001.1 | XP_021111660.1 | 3 - 4 | 247 | 110 | ||
| XM_021256002.1 | XP_021111661.1 | 3 - 4 | 765 | 110 | ||
| XM_021256003.1 | XP_021111662.1 | 3 - 4 | 437 | 110 | ||
More Information
Set Membership: |
|

Il7
Genomic Map