Assay Details
| Gene Symbol: LOC101716694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256352.1 | XP_021112011.1 | 1 - 2 | 72 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021256352.1 transcripts.
Set Membership: |
|

LOC101716694
Genomic Map