Assay Details
| Gene Symbol: Ccnjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836454.2 | XP_004836511.1 | 2 - 3 | 248 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccnjl| Gene Symbol: Ccnjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836454.2 | XP_004836511.1 | 2 - 3 | 248 | 82 | |
Set Membership: |
|