Assay Details
| Gene Symbol: Ccnjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836454.2 | XP_004836511.1 | 4 - 5 | 761 | 75 | |
| XM_004836455.2 | XP_004836512.1 | 3 - 4 | 657 | 75 | ||
| XM_021240280.1 | XP_021095939.1 | 4 - 5 | 808 | 75 | ||
More Information
Set Membership: |
|

Ccnjl
Genomic Map