Assay Details
| Gene Symbol: Ube2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839938.3 | XP_004839995.1 | 6 - 7 | 527 | 66 | |
| XM_004839939.3 | XP_004839996.1 | 6 - 7 | 463 | 66 | ||
| XM_021254303.1 | XP_021109962.1 | 7 - 8 | 722 | 66 | ||
| XM_021254304.1 | XP_021109963.1 | 7 - 8 | 642 | 66 | ||
| XM_021254307.1 | XP_021109966.1 | 5 - 6 | 419 | 66 | ||
More Information
Set Membership: |
|

Ube2d4
Genomic Map