Assay Details
| Gene Symbol: Lime1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841086.3 | XP_004841143.1 | 1 - 2 | 1284 | 75 | |
| XM_021255280.1 | XP_021110939.1 | 2 - 3 | 533 | 75 | ||
More Information
Set Membership: |
|

Lime1
Genomic Map