Assay Details
| Gene Symbol: Lime1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841086.3 | XP_004841143.1 | 3 - 4 | 1906 | 69 | |
| XM_021255280.1 | XP_021110939.1 | 4 - 5 | 1155 | 69 | ||
| XM_021255281.1 | XP_021110940.1 | 3 - 4 | 591 | 69 | ||
More Information
Set Membership: |
|

Lime1
Genomic Map