Assay Details
| Gene Symbol: Gprc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845854.2 | XP_004845911.2 | 2 - 3 | 1161 | 83 | |
| XM_021259162.1 | XP_021114821.1 | 3 - 4 | 1599 | 83 | ||
More Information
Set Membership: |
|

Gprc5d
Genomic Map