Assay Details
| Gene Symbol: LOC101726751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847765.3 | XP_004847822.1 | 2 - 3 | 367 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101726751| Gene Symbol: LOC101726751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847765.3 | XP_004847822.1 | 2 - 3 | 367 | 67 | |
Set Membership: |
|