Assay Details
| Gene Symbol: Htatip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851555.2 | XP_004851612.1 | 1 - 2 | 134 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htatip2| Gene Symbol: Htatip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851555.2 | XP_004851612.1 | 1 - 2 | 134 | 97 | |
Set Membership: |
|