Assay Details
| Gene Symbol: Mylk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873886.2 | XP_004873943.1 | 7 - 8 | 1945 | 63 | |
| XM_004873887.2 | XP_004873944.1 | 7 - 8 | 1559 | 63 | ||
| XM_004873888.2 | XP_004873945.1 | 7 - 8 | 1624 | 63 | ||
| XM_021251477.1 | XP_021107136.1 | 5 - 6 | 936 | 63 | ||
| XM_021251478.1 | XP_021107137.1 | 5 - 6 | 940 | 63 | ||
More Information
Set Membership: |
|

Mylk3
Genomic Map