Assay Details
| Gene Symbol: Utp14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004921148.3 | XP_004921205.2 | 2 - 3 | 257 | 72 | |
| XM_013077904.2 | XP_012933358.1 | 2 - 3 | 257 | 72 | ||
More Information
Set Membership: |
|

Utp14a
Genomic Map