Assay Details
| Gene Symbol: C1rl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067294.2 | XP_012922748.1 | 4 - 5 | 781 | 82 | |
| XM_013067296.2 | XP_012922750.1 | 4 - 5 | 780 | 82 | ||
| XM_013067297.2 | XP_012922751.1 | 4 - 5 | 770 | 82 | ||
More Information
Set Membership: |
|

C1rl
Genomic Map