Assay Details
Gene Symbol: Dupd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013072791.2 | XP_012928245.1 | 1 - 2 | 132 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013072791.2 transcripts.
Set Membership: |
![]() ![]() |
