Assay Details
| Gene Symbol: Dnlz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073871.2 | XP_012929325.1 | 2 - 3 | 455 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnlz| Gene Symbol: Dnlz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073871.2 | XP_012929325.1 | 2 - 3 | 455 | 100 | |
Set Membership: |
|