Assay Details
| Gene Symbol: LOC110345227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242034.1 | XP_021097693.1 | 1 - 2 | 44 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110345227| Gene Symbol: LOC110345227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242034.1 | XP_021097693.1 | 1 - 2 | 44 | 119 | |
Set Membership: |
|