Assay Details
| Gene Symbol: Zim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245706.1 | XP_021101365.1 | 2 - 3 | 207 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021245706.1 transcripts.
Set Membership: |
|

Zim2
Genomic Map