Assay Details
| Gene Symbol: LOC106008944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021254099.1 | XP_021109758.1 | 3 - 4 | 476 | 87 | |
| XM_021254100.1 | XP_021109759.1 | 3 - 4 | 316 | 87 | ||
More Information
Set Membership: |
|

LOC106008944
Genomic Map