Assay Details
| Gene Symbol: Extl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850631.3 | XP_004850688.1 | 7 - 8 | 2477 | 70 | |
| XM_021263095.1 | XP_021118754.1 | 7 - 8 | 2477 | 70 | ||
| XM_021263096.1 | XP_021118755.1 | 7 - 8 | 4236 | 70 | ||
More Information
Set Membership: |
|

Extl1
Genomic Map