Assay Details
| Gene Symbol: Kctd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853136.2 | XP_004853193.1 | 2 - 3 | 2270 | 72 | |
| XM_013075301.2 | XP_012930755.1 | 2 - 3 | 291 | 72 | ||
| XM_013075304.2 | XP_012930758.1 | 2 - 3 | 330 | 72 | ||
More Information
Set Membership: |
|

Kctd1
Genomic Map