Assay Details
| Gene Symbol: Wisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858183.2 | XP_004858240.1 | 4 - 5 | 1000 | 84 | |
| XM_021239078.1 | XP_021094737.1 | 4 - 5 | 687 | 84 | ||
| XM_021239079.1 | XP_021094738.1 | 4 - 5 | 994 | 84 | ||
More Information
Set Membership: |
|

Wisp2
Genomic Map