Assay Details
| Gene Symbol: Sertad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874203.2 | XP_004874260.1 | 1 - 2 | 153 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004874203.2 transcripts.
Set Membership: |
|

Sertad1
Genomic Map