Assay Details
| Gene Symbol: Ptgdr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874436.2 | XP_004874493.2 | 1 - 2 | 1879 | 71 | |
| XM_021251853.1 | XP_021107512.1 | 2 - 3 | 1738 | 71 | ||
More Information
Set Membership: |
|

Ptgdr2
Genomic Map